Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCRL5/FCRL3 Antibody (307307), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB20871APCCY7
Description
FCRL5/FCRL3 Monoclonal antibody specifically detects FCRL5/FCRL3 in Human samples. It is validated for Flow CytometrySpecifications
FCRL5/FCRL3 | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Mouse myeloma cell line NS0-derived recombinant human FCRL5/FCRL3, Gln16-Arg844, Accession # AAI01067 | |
0.1 mL | |
Primary | |
Store at 4C in the dark. Do not freeze. | |
IgG2b |
Flow Cytometry | |
307307 | |
Flow Cytometry | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction