Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCRL5 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 265031AP150UL

Description
This gene encodes a member of the immunoglobulin receptor superfamily and the Fc-receptor like family. This gene and several other Fc receptor-like gene members are clustered on the long arm of chromosome 1. The encoded protein is a single-pass type I membrane protein and contains 8 immunoglobulin-like C2-type domains. This gene is implicated in B cell development and lymphomagenesis. Alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
FCRL5 | |
Polyclonal | |
Unconjugated | |
FCRL5 | |
BXMAS1, CD307, Fc receptor homolog 5, Fc receptor like 5, Fc receptor like protein 5, FcR like protein 5, FCRH5, FCRL5, IRTA2, PRO820 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
83416 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96RD9 | |
FCRL5 | |
FCRL5 Fusion Protein Ag24351 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction