Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCRL6/FcRH6 Antibody (672505), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6745APCCY7
Description
FCRL6/FcRH6 Monoclonal antibody specifically detects FCRL6/FcRH6 in Human samples. It is validated for Flow CytometrySpecifications
FCRL6/FcRH6 | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
343413 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Flow Cytometry | |
672505 | |
Flow Cytometry | |
Fc receptor-like 6, FLJ16056 | |
Chinese hamster ovary cell line CHO-derived recombinant human FCRL6/FcRH6 , Lys16-Leu312, Accession # Q6DN72 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction