Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCRL6/FcRH6 Antibody (672505) [Janelia Fluor™ 646], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6745J
Description
FCRL6/FcRH6 Monoclonal antibody specifically detects FCRL6/FcRH6 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
FCRL6/FcRH6 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
343413 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, CyTOF | |
672505 | |
Flow Cytometry, CyTOF-ready | |
Fc receptor-like 6, FLJ16056 | |
Chinese hamster ovary cell line CHO-derived recombinant human FCRL6/FcRH6 , Lys16-Leu312, Accession # Q6DN72 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction