Learn More
Invitrogen™ FCRL6 Monoclonal Antibody (672505)

Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Specifications
Specifications
| Antigen | FCRL6 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 672505 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | FCRL6 |
| Gene Accession No. | Q6DN72 |
| Gene Alias | Fc receptor homolog 6; Fc receptor like 6; Fc receptor-like 6; Fc receptor-like protein 6; Fc receptor-like protein 7; FCRH6; FCRL6; fcR-like protein 6; FLJ16056; IFGP6; IgSF type I transmembrane receptor; leukocyte receptor |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.