Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCRL6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112982
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112982 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112982 Supplier Invitrogen™ Supplier No. PA5112982
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FCRL6 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Acts as a MHC class II receptor (PubMed:20519654). When stimulated on its own, does not play a role in cytokine production or the release of cytotoxic granules by NK cells and cytotoxic CD8(+) T cells (PubMed:17213291, PubMed:18991291). Does not act as an Fc receptor (PubMed:18991291). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCRL6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FCRL6
Gene Accession No. Q6DN72
Gene Alias Fc receptor homolog 6; Fc receptor like 6; Fc receptor-like 6; Fc receptor-like protein 6; Fc receptor-like protein 7; FCRH6; FCRL6; fcR-like protein 6; FLJ16056; IFGP6; IgSF type I transmembrane receptor; leukocyte receptor
Gene Symbols FCRL6
Host Species Rabbit
Immunogen Recombinant Human Fc receptor-like protein 6 protein (20-307AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 343413
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.