Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCRLA/FCRLM1 Antibody (865910) [DyLight 405], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB7905E
Description
FCRLA/FCRLM1 Monoclonal antibody specifically detects FCRLA/FCRLM1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
FCRLA/FCRLM1 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
84824 | |
Store at 4°C in the dark. | |
IgG2a |
Western Blot, Flow Cytometry, CyTOF | |
865910 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
Fc receptor homolog expressed in B cells (FREB), Fc receptor homolog expressed in B-cells, Fc receptor related protein X, Fc receptor-like A, Fc receptor-like and mucin-like 1, Fc receptor-related protein X, FCRL1, FCRLa, FCRLb, FCRLc1, FCRLc2, FCRLd, FCRLe, FCRLFc receptor-like protein, FCRLM1, FCRLX, FCRX, FREBFc receptor-like and mucin-like protein 1, MGC4595 | |
E. coli-derived recombinant mouse FCRLA, Lys221-Lys352, Accession # Q920A9 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction