Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCRLA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530731
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530731 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530731 Supplier Invitrogen™ Supplier No. PA530731
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Raji. Predicted reactivity: Human (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Receptors for the Fc fragment of IgG, or FCGRs (see MIM 146790), are cell surface glycoproteins of the Ig superfamily (IgSF). These receptors mediate phagocytosis of IgG-coated pathogens and promote activation of effector cells, leading to inflammatory responses and antibody-mediated cellular cytotoxicity. All FCGR genes map to human chromosome 1. Additional genes in this region, including FREB, encode FCGR homologs that are selectively expressed in B cells and may be implicated in B-cell development and lymphomagenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCRLA
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene FCRLA
Gene Accession No. Q7L513
Gene Alias Fc receptor homolog expressed in B cells (FREB); fc receptor homolog expressed in B-cells; Fc receptor like A; Fc receptor related protein X; Fc receptor-like A; Fc receptor-like and mucin-like 1; fc receptor-like and mucin-like protein 1; Fc receptor-like protein; fc receptor-related protein X; FCRL; FCRL1; FCRLA; FCRLb; FCRLc1; FCRLc2; FCRLd; FCRLe; FCRLM1; FCRLX; FCRX; FREB; UNQ291/PRO329
Gene Symbols FCRLA
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FCRLM1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84824
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.