Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCRLB/FCRY Antibody (454217), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4705E
Description
FCRLB/FCRY Monoclonal antibody specifically detects FCRLB/FCRY in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
FCRLB/FCRY | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
127943 | |
Store at 4°C in the dark. | |
IgG2b |
Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
454217 | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
Fc receptor homolog expressed in B cells protein 2, Fc receptor like 2, Fc receptor-like and mucin-like 2, Fc receptor-like and mucin-like protein 2, Fc receptor-like B, Fc receptor-like protein 2, Fc receptor-related protein Y, FCRL2FCRLM2, FCRLY, FCRY, FLJ31052, FREB2, FREB-2FcRY, RP11-474I16.6 | |
Mouse myeloma cell line NS0-derived recombinant human FCRLB/FCRY, Ala18-Ser426, Accession # Q6BAA4 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction