Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCRLB/FCRY Antibody (OTI5H6), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272384AF532
Description
FCRLB/FCRY Monoclonal specifically detects FCRLB/FCRY in Human, Rat samples. It is validated for Western Blot.Specifications
FCRLB/FCRY | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
FCRLB | |
Recombinant protein fragment corresponding to amino acids 202-426 of human FCRLB/FCRY (NP_001002901) produced in E.coli. | |
Primary | |
Human, Rat | |
IgG2b |
Western Blot | |
OTI5H6 | |
Western Blot | |
Fc receptor homolog expressed in B cells protein 2, Fc receptor like 2, Fc receptor-like and mucin-like 2, Fc receptor-like and mucin-like protein 2, Fc receptor-like B, Fc receptor-like protein 2, Fc receptor-related protein Y, FCRL2FCRLM2, FCRLY, FCRY, FLJ31052, FREB2, FREB-2FcRY, RP11-474I16.6 | |
Mouse | |
0.1 mL | |
127943 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction