Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCRLB/FCRY Antibody (OTI5H6), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245639
Description
Ensure accurate, reproducible results in Western Blot
FCRLB/FCRY Monoclonal specifically detects FCRLB/FCRY in Human, Rat samples. It is validated for Western Blot.Specifications
FCRLB/FCRY | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000 | |
Fc receptor homolog expressed in B cells protein 2, Fc receptor like 2, Fc receptor-like and mucin-like 2, Fc receptor-like and mucin-like protein 2, Fc receptor-like B, Fc receptor-like protein 2, Fc receptor-related protein Y, FCRL2FCRLM2, FCRLY, FCRY, FLJ31052, FREB2, FREB-2FcRY, RP11-474I16.6 | |
Mouse | |
Immunogen affinity purified | |
RUO | |
127943 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2b |
Western Blot | |
OTI5H6 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
FCRLB | |
Human recombinant protein fragment corresponding to amino acids 202-426 of human FCRLB(NP_001002901) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction