Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FCRLB Monoclonal Antibody (454217)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524172
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
FCRL2 belongs to the Fc receptor family. Fc receptors are involved in phagocytosis, antibody-dependent cell cytotoxicity, immediate hypersensitivity, and transcytosis of immunoglobulins via their ability to bind immunoglobulin (Ig) constant regions.
Specifications
FCRLB | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q6BAA4 | |
FCRLB | |
Mouse myeloma cell line NS0-derived recombinant human FCRLB/FCRY Ala18-Ser426. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Western Blot | |
454217 | |
Unconjugated | |
FCRLB | |
fc receptor homolog expressed in B cells protein 2; Fc receptor homolog expressed in B-cells protein 2; Fc receptor like B; Fc receptor related Y; fc receptor-like and mucin-like protein 2; Fc receptor-like B; Fc receptor-like mucin-like 2; fc receptor-like protein 2; Fc receptor-related protein Y; FCRL2; FCRL2FCRLM2; FCRLB; FCRLM2; FCRLY; FcRY; FLJ31052; FREB2; FREB-2; FREB-2FcRY; mFCRL2; RGD1563018; RP11-474I16.6 | |
Mouse | |
Protein A/G | |
RUO | |
127943 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction