Learn More
Invitrogen™ FCRLB Monoclonal Antibody (OTI4C4)

Description
FCRLB Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | FCRLB |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI4C4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | FCRLB |
| Gene Accession No. | Q6BAA4 |
| Gene Alias | fc receptor homolog expressed in B cells protein 2; Fc receptor homolog expressed in B-cells protein 2; Fc receptor like B; Fc receptor related Y; fc receptor-like and mucin-like protein 2; Fc receptor-like B; Fc receptor-like mucin-like 2; fc receptor-like protein 2; Fc receptor-related protein Y; FCRL2; FCRL2FCRLM2; FCRLB; FCRLM2; FCRLY; FcRY; FLJ31052; FREB2; FREB-2; FREB-2FcRY; mFCRL2; RGD1563018; RP11-474I16.6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.