Learn More
FCRLB Mouse anti-Human, Clone: OTI4G1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809247

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
FCRL2 belongs to the Fc receptor family. Fc receptors are involved in phagocytosis, antibody-dependent cell cytotoxicity, immediate hypersensitivity, and transcytosis of immunoglobulins via their ability to bind immunoglobulin (Ig) constant regions.Specifications
FCRLB | |
Monoclonal | |
Unconjugated | |
FCRLB | |
FCRL2, FCRLM2, FCRLY, FREB-2, FREB2, FcRY | |
Mouse | |
Affinity Chromatography | |
RUO | |
127943 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4G1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q6BAA4 | |
FCRLB | |
Human recombit protein fragment corresponding to amino acids 202-426 of human FCRLB produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.