Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCRN/FCGRT Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$416.50 - $706.00
Specifications
Antigen | FCRN/FCGRT |
---|---|
Dilution | Western Blot 0.04 - 0.4 ug/ml, Simple Western 1:20, Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:5000 - 1:10000, Immunohistochemistry-Frozen Reported in scientific literature (PMID 24278022)., Knockout Validated |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
FCRN/FCGRT Polyclonal specifically detects FCRN/FCGRT in Human, Cynomolgus Monkey samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.Specifications
FCRN/FCGRT | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
RUO | |
Human, Cynomolgus Monkey | |
P55899 | |
2217 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LTFLLFSCPHRLREHLERGRGNLEWKEPPSMRLKARPSSPGFSVLTCSAFSFYPPELQLRFLRNGLAAGTGQGDFGPNSDGSFHASSSLTVKSGDEHHYCCIVQHAGLAQPLRVELESPAKS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
40 kDa |
Western Blot 0.04 - 0.4 ug/ml, Simple Western 1:20, Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:5000 - 1:10000, Immunohistochemistry-Frozen Reported in scientific literature (PMID 24278022)., Knockout Validated | |
Polyclonal | |
Rabbit | |
Immunology | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
alpha-chain, Fc fragment of IgG, receptor, transporter, alpha, FcRn, FcRn alpha chain, FCRNimmunoglobulin receptor, intestinal, heavy chain, IgG Fc fragment receptor transporter alpha chain, IgG receptor FcRn large subunit p51, major histocompatibility complex class I-like Fc receptor, Neonatal Fc receptor, neonatal Fc-receptor for Ig | |
FCGRT | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title