Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCRN/FCGRT Mouse anti-Human, Clone: FCGRT/2932, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | FCRN/FCGRT |
---|---|
Clone | FCGRT/2932 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 2-4 ug/ml, Protein Array |
Applications | Immunohistochemistry (Paraffin), Peptide Array |
Description
FCRN/FCGRT Monoclonal specifically detects FCRN/FCGRT in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
FCRN/FCGRT | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Human | |
alpha-chain, Fc fragment of IgG, receptor, transporter, alpha, FcRn, FcRn alpha chain, FCRNimmunoglobulin receptor, intestinal, heavy chain, IgG Fc fragment receptor transporter alpha chain, IgG receptor FcRn large subunit p51, major histocompatibility complex class I-like Fc receptor, Neonatal Fc receptor, neonatal Fc-receptor for Ig | |
Recombinant fragment of human FCRN/FCGRT protein (around aa 24-215) (Exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
FCGRT/2932 | |
Immunohistochemistry-Paraffin 2-4 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
2217 | |
IgG2b κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title