Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FcRn (FCGRT) Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABF19125UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABF191-25UL 25 μL
ABF191MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABF191-25UL Supplier MilliporeSigma Supplier No. ABF19125UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This rabbit polyclonal Anti-FcRn (FCGRT) antibody detects IgG receptor FcRn large subunit p51. It has been tested for use in Immunofluorescence and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FcRn (FCGRT)
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Fcgrt,Fcrn
Host Species Rabbit
Immunogen GST-tagged recombinant fragment corresponding to 44 amino acids from the C-terminal region of murine IgG receptor FcRn large subunit p51.
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity This polyclonal antibody detects murine IgG receptor FcRn. It targets an epitope within 44 amino acids from the C-terminal region.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.