Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCGRT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579245
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579245 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579245 Supplier Invitrogen™ Supplier No. PA579245
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: A549 whole cell. IHC: human lung cancer tissue.

FCGRT binds to the Fc region of monomeric immunoglobulins gamma. It mediates the uptake of IgG from milk. It plays a possible role in transfer of immunoglobulin G from mother to fetus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCGRT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene FCGRT
Gene Accession No. P55899
Gene Alias alpha-chain; Fc fragment of IgG receptor and transporter; Fc fragment of IgG, receptor, transporter, alpha; Fc receptor, IgG, alpha chain transporter; FCGRT; FCRN; FcRn alpha chain; heavy chain of the major histocompatibility complex class I-like Fc receptor; IgG Fc fragment receptor transporter alpha chain; IgG receptor FcRn large subunit p51; immunoglobulin receptor, intestinal, heavy chain; major histocompatibility complex class I-like Fc receptor; neonatal Fc receptor; neonatal Fc-receptor for Ig; transmembrane alpha chain of the neonatal receptor
Gene Symbols FCGRT
Host Species Rabbit
Immunogen E. coli-derived human FCGRT recombinant protein (Position: Q56-E291).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2217
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.