Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FD and C Blue No. 1, Spectrum™ Chemical

Click to view available options
Quantity:
25 g
100 g
500 g
10 kg
45 kg
Packaging:
Fiber Drum
Description
Spectrum™ Chemical FD and C Blue No. 1 is a synthetic dye produced from petroleum. It is used as a coloring agent, biological stain, textile dye, wood stain, and indicator. In cosmetics and personal care products, it is used in the formulation of a wide variety of product types, eye makeup, makeup, lipsticks, nail polishes, personal cleansers and skin care products. It appears as a dark violet to reddish violet, crystalline powder that is water soluble. In the United States, the Food and Drug Administration (FDA) controls the specifications for food, drug and cosmetic colors. Spectrum certifies all batches of artificial food colors meet FDA FD&C quality.
Specifications
Specifications
CAS Max % | 100% |
Percent Purity | 85% |
Grade | Reagent |
Packaging | Fiber Drum |
Color | Purple/Violet |
Quantity | 500 g |
Chemical Name or Material | FD and C Blue No. 1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction