Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FD and C Red No. 3, Spectrum™ Chemical
SDP

Catalog No. 18602627 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
20 kg
Packaging:
Amber Glass Bottle

FD135, 16423-68-0, C20H6O5I4Na2

Spectrum™ Chemical FD and C Red No. 3 is used as a food, drug and cosmetic dye and originally used as a replacement for amaranth as a food coloring. It looks like a dark red powder and often comes as a sodium salt. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 16423-68-0
Molecular Formula C20H6I4Na2O5
Molecular Weight (g/mol) 879.86
MDL Number MFCD00144257
InChI Key IINNWAYUJNWZRM-UHFFFAOYSA-L
IUPAC Name disodium 2-(2,4,5,7-tetraiodo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=C(C=CC=C1)C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12

Specifications

CAS Max % 100%
Grade Reagent
Packaging Amber Glass Bottle
Color Maroon
Quantity 100 g
Chemical Name or Material FD and C Red No. 3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.