Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FD and C Red No. 3, Spectrum™ Chemical

Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
20 kg
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical FD and C Red No. 3 is used as a food, drug and cosmetic dye and originally used as a replacement for amaranth as a food coloring. It looks like a dark red powder and often comes as a sodium salt. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
CAS Max % | 100% |
Grade | Reagent |
Packaging | Amber Glass Bottle |
Color | Maroon |
Quantity | 5 kg |
Chemical Name or Material | FD and C Red No. 3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction