Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
fd/M13 bacteriophage Antibody (MM05), DyLight 488 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305788G
Description
fd/M13 bacteriophage Monoclonal antibody specifically detects fd/M13 bacteriophage in Virus samples. It is validated for ELISASpecifications
fd/M13 bacteriophage | |
Monoclonal | |
DyLight 488 | |
major coat protein | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with M13 Bacteriophage. | |
0.1 mL | |
Primary | |
Virus | |
Purified |
ELISA | |
MM05 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
927333 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction