Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

fd/M13 bacteriophage Antibody, Novus Biologicals™
SDP

Catalog No. p-5071210 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001633SS 0.02 mL
NB1001633 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1001633SS Supplier Novus Biologicals Supplier No. NB1001633SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 5 publications

fd/M13 bacteriophage Polyclonal specifically detects fd/M13 bacteriophage in Virus samples. It is validated for Flow Cytometry, ELISA, Lateral Flow Assay.

Specifications

Antigen fd/M13 bacteriophage
Applications ELISA
Classification Polyclonal
Concentration 4.3 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:1000 (indirect)
Formulation 0.01 M phosphate buffered saline, pH 7.4 with 15mM Sodium Azide
Gene Accession No. P69541
Gene Alias major coat protein
Gene Symbols VIII
Host Species Rabbit
Immunogen Repeated injections with fd bacteriophage.
Purification Method IgG purified
Quantity 0.02 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 927333
Test Specificity The antibody binds specifically to phage coat proteins of fd phage or M13 phage.
Target Species Virus
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.