Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ FDAA (Marfey's Reagent) (1-fluoro-2-4-dinitrophenyl-5-L-alanine amide) PROMO

Catalog No. PI48895PM
Click to view available options
:
Thermo Scientific™ FDAA (Marfey's Reagent) (1-fluoro-2-4-dinitrophenyl-5-L-alanine amide)

Catalog No. PI48895PM

Supplier: Thermo Scientific™ 48895

Only null left
Add to Cart
Add to Cart

Label and measure optical isomers of amino acids by reverse-phase chromatography (HPLC) with this amine-reactive, colored (Amax 340nm) compound.

Highlights:

  • Optical isomers of amino acids derivatization complete in 90 minutes
  • Derivatives have an absorption coefficient of ∽3 x 104 Derivatives can be detected by UV at 340nm with picomole sensitivity

Specifications

Product Type FDAA
Chemical Reactivity Amine
Content And Storage Upon receipt store product at room temperature.
Label or Dye Other Label or Dye
Quantity 50 mg
Form Solid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.