Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FDC Antibody (CNA.42), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | FDC |
---|---|
Clone | CNA.42 |
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP14311803
![]() |
Novus Biologicals
NBP1431180.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NBP14311801
![]() |
Novus Biologicals
NBP1431180.1MG |
0.1 mg |
Each for $499.50
|
|
|||||
Description
FDC Monoclonal specifically detects FDC in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
FDC | |
0.5 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
RUO | |
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
1218 | |
The immunogen for this antibody was follicular dendritic cells. | |
Primary | |
Store at 4C. Do not freeze. |
CNA.42 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Mouse | |
Human | |
cardiomyopathy, dilated 1B (autosomal dominant), CMPD1, FDC | |
CMD1B | |
IgM | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title