Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FDCSP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599876
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599876 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599876 Supplier Invitrogen™ Supplier No. PA599876
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FDCSP.

This gene encodes a small secreted protein that is expressed in follicular dendritic cells. This protein specifically binds to activated B cells, and functions as a regulator of antibody responses. It is also thought to contribute to tumor metastases by promoting cancer cell migration and invasion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FDCSP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FDCSP
Gene Accession No. Q8NFU4
Gene Alias C4orf7; FDC secreted protein; FDCSP; FDC-SP; follicular dendritic cell secreted; follicular dendritic cell secreted peptide; follicular dendritic cell secreted protein; LOC494224; UNQ733/PRO1419
Gene Symbols FDCSP
Host Species Rabbit
Immunogen A synthesized peptide derived from human FDCSP(Accession Q8NFU4), corresponding to amino acid residues R26-A76.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 260436
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.