Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FDFT1 Antibody (OTI2F10), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP27071A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27071A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27071A32 Supplier Novus Biologicals Supplier No. NBP270715AF532
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FDFT1 Monoclonal specifically detects FDFT1 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FDFT1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2F10
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DGPT, EC 2.5.1.21, ERG9, Farnesyl-diphosphate farnesyltransferase, farnesyl-diphosphate farnesyltransferase 1, FPP:FPP farnesyltransferase, presqualene-di-diphosphate synthase, SQS, squalene synthase, squalene synthetase, SS
Gene Symbols FDFT1
Host Species Mouse
Immunogen Full length recombinant protein of human FDFT1 (NP_004453) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2222
Target Species Human, Mouse, Rat, Monkey, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.