Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FDFT1 Recombinant Rabbit Monoclonal Antibody (10H12)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA538387
Description
This antibody has been tested in direct-ELISA.
FDFT1 catalyzes the first step in the cholesterol biosynthetic pathway, the conversion of trans-farnesyldiphosphate to squalene. The loss of promoter activity and response to sterols for FDFT1 is localized to a 69-bp section positioned 131 bp 5-prime to the transcription start site. Sequence analysis of this region shows that it contains a sterol regulatory element-1 (SRE1) previously identified in other sterol regulated genes and 2 putative NF1 binding sites.
Specifications
FDFT1 | |
Recombinant Monoclonal | |
1.5 mg/mL | |
PBS with 50% glycerol, 150 mM NaCl and 0.02% sodium azide; pH 7.4 | |
P37268 | |
FDFT1 | |
A synthesized peptide derived from human FDFT1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
10H12 | |
Unconjugated | |
FDFT1 | |
DGPT; EGK_18680; ERG9; farnesyl diphosphate farnesyl transferase 1; farnesyl diphosphate farnesyltransferase; farnesyl-diphosphate farnesyltransferase; farnesyl-diphosphate farnesyltransferase 1; FDFT1; FPP:FPP farnesyltransferase; presqualene-di-diphosphate synthase; SQS; Squalene synthase; squalene synthetase; SS | |
Rabbit | |
Affinity chromatography | |
RUO | |
2222 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction