Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FDFT1 Rabbit anti-Human, Mouse, Rat, Clone: 4W10J4, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | FDFT1 |
---|---|
Clone | 4W10J4 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
FDFT1 Monoclonal antibody specifically detects FDFT1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
FDFT1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Cardiovascular Biology, Endocrinology, Neuroscience | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
2222 | |
IgG | |
Affinity purified |
4W10J4 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
DGPT, EC 2.5.1.21, ERG9, Farnesyl-diphosphate farnesyltransferase, farnesyl-diphosphate farnesyltransferase 1, FPP:FPP farnesyltransferase, presqualene-di-diphosphate synthase, SQS, squalene synthase, squalene synthetase, SS | |
A synthetic peptide corresponding to a sequence within amino acids 150-250 of human FDFT1 (P37268). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title