Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FDPS Antibody (115), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290812AF750
Description
FDPS Monoclonal specifically detects FDPS in Mouse samples. It is validated for ELISA.Specifications
FDPS | |
Monoclonal | |
Alexa Fluor 750 | |
6E)-farnesyl diphosphate synthase, Dimethylallyltranstransferase, EC 2.5.1.1, EC 2.5.1.10, farnesyl diphosphate synthaseFPP synthetase, farnesyl pyrophosphate synthase, farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, FPP synthase, FPPS, FPSgeranyltranstransferase, Geranyltranstransferase, KIAA1293 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse FDPS (NP_608219.1; Met1-Lys353). | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
2224 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
115 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction