Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FDPS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596411
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596411 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596411 Supplier Invitrogen™ Supplier No. PA596411
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IGGKYNRGLT VVVAFRELVE PRKQDADSLQ RAWTVGWCVE LLQAFFLVAD DIMDSSLTRR GQICWYQKPG VGLDAINDAN LLEACIYRLL KLYCREQPYY LNLIELFLQS SYQTEIGQTL DLLTAPQGNV DLVRFTEKRY KSIVKYKTAF YSFYLPIAAA MYMAGIDGEK EHANAKKILL EMGEFFQIQD DYLDLFGDPS VTGKIGTDIQ DNKCSWLVVQ CLQRATPEQY QILKENYGQK EAEKVARVKA LYEELDLPAV FLQYEEDSYS HIMALIEQYA APLPPAVFLG LARKIYKRRK; Positive Samples: 293T, HepG2, HeLa, MCF7, U-251MG, BxPC-3, Mouse liver, Mouse ovary, Rat liver, Rat testis; Cellular Location: Cytoplasm.

The isoprene biosynthetic pathway supply the cell with cholesterol, ubiquinone, and various nonsterol metabolites. The farnesylpyrophosphate synthetase enzyme catalyzes the formation of geranyl and farnesylpyrophosphate from isopentenylpyrophosphate and dimethylallyl pyrophosphate. Analysis of FDPS activity and protein in rat liver, accompanied by immunofluorescence and immunoelectron microscopy studies, demonstrated that FDPS is predominantly localized in peroxisomes. 1 Liver tissue from patients with the peroxisomal deficiency diseases Zellweger syndrome and neonatal adrenoleukodystrophy exhibit diminished activities of FDPS and subsequent isoprenoid synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FDPS
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FDPS
Gene Accession No. P05369, P14324, Q920E5
Gene Alias (2E,6E)-farnesyl diphosphate synthase; 6030492I17Rik; Ac2-125; AI256750; cholesterol-regulated 39 kDa protein; CR 39; dimethylallyltranstransferase; Farnesyl diphosphate synthase; farnesyl diphosphate synthase (farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, geranyltranstransferase); farnesyl diphosphate synthetase; farnesyl pyrophosphate synthase; farnesyl pyrophosphate synthetase; farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, geranyltranstransferase; Fdps; Fdpsl1; FPP synthase; FPP synthetase; FPPS; FPS; geranyltranstransferase; KIAA1293; mKIAA1293; POROK9; testis-specific farnesyl pyrophosphate synthetase
Gene Symbols FDPS
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 120-419 of human FPS/FDPS (NP_0019951).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110196, 2224, 83791
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.