Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FDPS Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982657
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2657 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2657 Supplier Bioss Supplier No. BS13153R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The isoprene biosynthetic pathway supply the cell with cholesterol, ubiquinone, and various nonsterol metabolites. The farnesylpyrophosphate synthetase enzyme catalyzes the formation of geranyl and farnesylpyrophosphate from isopentenylpyrophosphate and dimethylallyl pyrophosphate. Analysis of FDPS activity and protein in rat liver, accompanied by immunofluorescence and immunoelectron microscopy studies, demonstrated that FDPS is predominantly localized in peroxisomes. 1 Liver tissue from patients with the peroxisomal deficiency diseases Zellweger syndrome and neonatal adrenoleukodystrophy exhibit diminished activities of FDPS and subsequent isoprenoid synthesis.

Specifications

Antigen FDPS
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FDPS
Gene Accession No. P14324
Gene Alias (2E,6E)-farnesyl diphosphate synthase; 6030492I17Rik; Ac2-125; AI256750; Cholesterol-regulated 39 kDa protein; CR 39; Dimethylallyltranstransferase; Farnesyl diphosphate synthase; farnesyl diphosphate synthase (farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, geranyltranstransferase); farnesyl diphosphate synthetase; Farnesyl pyrophosphate synthase; farnesyl pyrophosphate synthetase; farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, geranyltranstransferase; Fdps; Fdpsl1; FPP synthase; FPP synthetase; FPPS; FPS; Geranyltranstransferase; KIAA1293; mKIAA1293; POROK9; testis-specific farnesyl pyrophosphate synthetase
Gene Symbols FDPS
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FDPS.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2224
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.