Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FDX1 Monoclonal Antibody (1E7)
GREENER_CHOICE

Catalog No. PIMA524558
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524558 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524558 Supplier Invitrogen™ Supplier No. MA524558
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: VGDSLLDVVV ENNLDIDGFG ACEGTLACST CHLIFEDHIY EKLDAITDEE NDMLDLAYGL TDRSRLGCQI CLTKSMDNMT VRVPETVADA RQSIDVGKT.

The product of this gene is a small iron-sulfur protein that transfers electrons from NADPH through ferredoxin reductase to a terminal cytochrome P450. This particular oxidation/reduction system is found in steroidogenic tissues, and is involved with the synthesis of bile acid and vitamin D. In addition to the expressed gene at this chromosomal locus (11q22), there are pseudogenes located on chromosomes 20 and 21. This gene product has been identified in a number of different tissues but all forms have been shown to be identical and are not tissue specific.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FDX1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene FDX1
Gene Accession No. P10109
Gene Alias adrenal ferredoxin; Adrenodoxin, mitochondrial; ADX; FDX; Fdx1; ferredoxin 1; ferredoxin-1; Hepatoredoxin; LOH11CR1D; mitochondrial adrenodoxin
Gene Symbols FDX1
Host Species Mouse
Immunogen FDX1 (NP_004100.1, 85 a.a. ~ 183 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2230
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.