Learn More
Invitrogen™ FDX1 Polyclonal Antibody

Description
Immunogen sequence: MAAAGGARLL RAASAVLGGP AGRWLHHAGS RAGSSGLLRN RGPGGSAEAS RSLSVSARAR SSSEDKITVH FINRDGETLT TKGKVGDSLL DVVVENNLDI DGFGACEGTL ACSTCHLIFE DHIYEKLDAI TDEENDMLDL AYGLTDRSRL GCQICLTKSM DNMTVRVPET VADARQSIDV GKTS; Positive Samples: 293T, K-562, DU145; Cellular Location: Mitochondrion matrix.
Specifications
Specifications
| Antigen | FDX1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | FDX1 |
| Gene Accession No. | P10109 |
| Gene Alias | adrenal ferredoxin; Adrenodoxin, mitochondrial; ADX; FDX; Fdx1; ferredoxin 1; ferredoxin-1; Hepatoredoxin; LOH11CR1D; mitochondrial adrenodoxin |
| Gene Symbols | FDX1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.