Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FDX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 125921AP150UL

Description
The product of this gene is a small iron-sulfur protein that transfers electrons from NADPH through ferredoxin reductase to a terminal cytochrome P450. This particular oxidation/reduction system is found in steroidogenic tissues, and is involved with the synthesis of bile acid and vitamin D. In addition to the expressed gene at this chromosomal locus (11q22), there are pseudogenes located on chromosomes 20 and 21. This gene product has been identified in a number of different tissues but all forms have been shown to be identical and are not tissue specific.Specifications
FDX1 | |
Polyclonal | |
Unconjugated | |
FDX1 | |
Adrenal ferredoxin, Adrenodoxin, mitochondrial, ADX, FDX, FDX1, ferredoxin 1, Hepatoredoxin, LOH11CR1D | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14148, 2230, 29189 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P10109, P24483, P46656 | |
Fdx1 | |
FDX1 Fusion Protein Ag3301 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction