Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FDXACB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558625
Description
Immunogen sequence: HPIKTINEKL IAELGKVFPL KRLKCSYPLL PQEGTSVLPF WNCDFLSAAF WISLHEDNSN SESLTGGTSQ DVEDFLVSFS ELSLLKNPGR DGKEEACEGT CG Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 57%.

Specifications
FDXACB1 | |
Polyclonal | |
Unconjugated | |
FDXACB1 | |
FDXACB1; FDX-ACDB domain-containing protein 1; ferredoxin-fold anticodon binding domain containing 1; ferredoxin-fold anticodon-binding domain-containing protein 1; hCG_2033039; hypothetical protein BC006136 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91893 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BRP7 | |
FDXACB1 | |
Recombinant protein corresponding to Human FDXACB1. Recombinant protein control fragment (Product #RP-97346). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction