Learn More
Invitrogen™ FDXR Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596260
Description
Immunogen sequence: PILDPVDFLG LQDKIKEVPR PRKRLTELLL RTATEKPGPA EAARQASASR AWGLRFFRSP QQVLPSPDGR RAAGVRLAVT RLEGVDEATR AVPTGDMEDL PCGLVLSSIG YKSRPVDPSV PFDSKLGVIP NVEGRVMDVP GLYCSGWVKR GPTGVIATTM TDSFLTGQML LQDLKAGLLP SGPRPGYAAI QALLSSRGVR PVSFSDWEKL DAEEVARGQG TGKPREKLVD PQEMLRLLGH; Positive Samples: HepG2, NCI-H460, Mouse kidney, Mouse testis, Mouse ovary, Rat lung, Rat kidney; Cellular Location: Mitochondrion matrix.
This gene encodes a member of the prefoldin beta subunit family. The encoded protein is one of six subunits of prefoldin, a molecular chaperone complex that binds and stabilizes newly synthesized polypeptides, thereby allowing them to fold correctly. The complex, consisting of two alpha and four beta subunits, forms a double beta barrel assembly with six protruding coiled-coils.
Specifications
FDXR | |
Polyclonal | |
Unconjugated | |
FDXR | |
adrenodoxin reductase; adrenodoxin-NADP(+) reductase; ADXR; AR; FDXR; Ferredoxin reductase; ferredoxin--NADP(+) reductase; NADPH:adrenodoxin oxidoreductase, mitochondrial | |
Rabbit | |
Affinity chromatography | |
RUO | |
14149, 2232, 79122 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.03 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P22570, P56522, Q61578 | |
FDXR | |
Recombinant fusion protein containing a sequence corresponding to amino acids 252-491 of human FDXR (NP_0777283). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.