Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FDXR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597743
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597743 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597743 Supplier Invitrogen™ Supplier No. PA597743
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FDXR Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a member of the prefoldin beta subunit family. The encoded protein is one of six subunits of prefoldin, a molecular chaperone complex that binds and stabilizes newly synthesized polypeptides, thereby allowing them to fold correctly. The complex, consisting of two alpha and four beta subunits, forms a double beta barrel assembly with six protruding coiled-coils.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FDXR
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FDXR
Gene Accession No. P22570, Q61578
Gene Alias adrenodoxin reductase; adrenodoxin-NADP(+) reductase; ADXR; AR; FDXR; Ferredoxin reductase; ferredoxin--NADP(+) reductase; NADPH:adrenodoxin oxidoreductase, mitochondrial
Gene Symbols FDXR
Host Species Rabbit
Immunogen Recombinant Human NADPH:adrenodoxin oxidoreductase, mitochondrial protein (33-451AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14149, 2232
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.