Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FE65 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11058360AF532
Description
FE65 Polyclonal specifically detects FE65 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
FE65 | |
Polyclonal | |
Western Blot | |
adaptor protein FE65a2, amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65), amyloid beta A4 precursor protein-binding, family B, member 1, Fe65, FE65amyloid beta A4 precursor protein-binding family B member 1, MGC:9072, Protein Fe65, RIR, stat-like protein | |
Rabbit | |
0.1 mL | |
322 | |
Store at 4°C in the dark. |
Western Blot | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
APBB1 | |
Recombinant rat FE65, expressed in bacteria. [Swiss-ProtNo. P46933] | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction