Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FE65 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11058360H
Description
FE65 Polyclonal specifically detects FE65 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
FE65 | |
Polyclonal | |
Western Blot | |
adaptor protein FE65a2, amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65), amyloid beta A4 precursor protein-binding, family B, member 1, Fe65, FE65amyloid beta A4 precursor protein-binding family B member 1, MGC:9072, Protein Fe65, RIR, stat-like protein | |
Rabbit | |
100 kDa | |
0.1 mL | |
Alzheimers Research, Cell Cycle and Replication, Neurodegeneration, Neuroscience, Signal Transduction | |
322 | |
Store at 4C in the dark. |
Western Blot | |
HRP | |
PBS with No Preservative | |
APBB1 | |
Recombinant rat FE65, expressed in bacteria. [Swiss-Prot# P46933] | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction