Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FE65 Antibody (SR1014), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322195100UL
Description
FE65 Monoclonal antibody specifically detects FE65 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
FE65 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
SR1014 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
adaptor protein FE65a2, amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65), amyloid beta A4 precursor protein-binding, family B, member 1, Fe65, FE65amyloid beta A4 precursor protein-binding family B member 1, MGC:9072, Protein Fe65, RIR, stat-like protein | |
A synthesized peptide derived from human FE65 (Uniprot #: O00213) | |
100 μg | |
Alzheimers Research, Cell Cycle and Replication, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction | |
322 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction