Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FE65 Mouse anti-Mouse, Human, Clone: 3H6, MilliporeSigma™

Catalog No. 05758I100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-758-I100UG 100 μg
05-758-I25UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-758-I100UG Supplier MilliporeSigma Supplier No. 05758I100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-FE65, clone 3H6 is a highly specific mouse monoclonal antibody that targets FE65 and has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FE65
Applications Western Blot
Classification Monoclonal
Clone 3H6
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Apbb1,FE65
Host Species Mouse
Immunogen GST-tgged recombinant fragment corresponding to 40 amino acids from the N-terminal half of murine Protein FE65.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001240814.1
Test Specificity Clone 3H6 specifically detects Protein FE65 in murine and human cells.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.