Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fe65 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518560
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518560 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518560 Supplier Invitrogen™ Supplier No. PA518560
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

The protein encoded by this gene is a member of the Fe65 protein family. It is an adaptor protein localized in the nucleus. It interacts with the Alzheimer's disease amyloid precursor protein, transcription factor CP2/LSF/LBP1 and the low-density lipoprotein receptor-related protein. APP functions as a cytosolic anchoring site that can prevent the gene product's nuclear translocation. This encoded protein could play an important role in the pathogenesis of Alzheimer's disease. It is thought to regulate transcription. Also it is observed to block cell cycle progression by downregulating thymidylate synthase expression. Multiple alternatively spliced transcript variants have been described for this gene but some of their full length sequence is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fe65
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene APBB1
Gene Accession No. Q9QXJ1
Gene Alias adaptor protein FE65a2; amyloid beta (A4) precursor protein-binding, family B, member 1; amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65); amyloid beta A4 precursor protein-binding family B member 1; amyloid beta precursor protein binding family B member 1; Amyloid-beta A4 precursor protein-binding family B member 1; APBB1; FE65; MGC:9072; Protein Fe65; RIR; stat-like protein
Gene Symbols APBB1
Host Species Goat
Immunogen Synthetic peptide sequence (GSLKPKRLGAHTP) corresponding to the C-terminus amino acids of APBB1 (aa 698-706).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11785
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.