Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FECH Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | FECH |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
FECH Polyclonal specifically detects FECH in Human, Mouse samples. It is validated for Western Blot.Specifications
FECH | |
Polyclonal | |
Rabbit | |
Lipid and Metabolism | |
EC 4.99.1.1, EPP, FCE, ferrochelatase, ferrochelatase (protoporphyria), ferrochelatase, mitochondrial, Heme synthase, heme synthetase, Protoheme ferro-lyase, protoporphyria | |
FECH | |
IgG | |
42 kDa |
Western Blot | |
Unconjugated | |
RUO | |
P22830 | |
2235 | |
Synthetic peptides corresponding to FECH(ferrochelatase (protoporphyria)) The peptide sequence was selected from the middle region of FECH. Peptide sequence KRSEVVILFSAHSLPMSVVNRGDPYPQEVSATVQKVMERLEYCNPYRLVW. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title