Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FECH Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
| Antigen | FECH |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15486320
![]() |
Novus Biologicals
NBP15486320UL |
20 μL |
Each for $206.00
|
|
|||||
NBP154863
![]() |
Novus Biologicals
NBP154863 |
100 μL |
Each for $487.50
|
|
|||||
Description
FECH Polyclonal specifically detects FECH in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FECH | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| EC 4.99.1.1, EPP, FCE, ferrochelatase, ferrochelatase (protoporphyria), ferrochelatase, mitochondrial, Heme synthase, heme synthetase, Protoheme ferro-lyase, protoporphyria | |
| FECH | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Q8NAN0 | |
| 2235 | |
| Synthetic peptides corresponding to FECH(ferrochelatase (protoporphyria)) The peptide sequence was selected from the N terminal of FECH. Peptide sequence QHAQGAKPQVQPQKRYESNIRKPKTGILMLNMGGPETLGDVHDFLLRLFL. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title