Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FECH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597044
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597044 Supplier Invitrogen™ Supplier No. PA597044
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MRSLGANMAA ALRAAGVLLR DPLASSSWRV CQPWRWKSGA AAAAVTTETA QHAQGAKPQV QPQKRKPKTG ILMLNMGGPE TLGDVHDFLL RLFLDRDLMT LPIQNKLAPF IAKRRTPKIQ EQYRRIGGGS PIKIWTSKQG EGMVKLLDEL SPNTAPHKYY IGFRYVHPLT EEAIEEMERD GLERAIAFTQ YPQYSCSTTG; Positive Samples: HepG2, HT-29, Mouse brain; Cellular Location: Matrix side, Mitochondrion inner membrane, Peripheral membrane protein.

The protein encoded by this gene is localized to the mitochondrion, where it catalyzes the insertion of the ferrous form of iron into protoporphyrin IX in the heme synthesis pathway. Mutations in this gene are associated with erythropoietic protoporphyria. Two transcript variants encoding different isoforms have been found for this gene. A pseudogene of this gene is found on chromosome 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FECH
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FECH
Gene Accession No. P22315, P22830
Gene Alias AI894116; EPP; FCE; fch; Fcl; FECH; ferrochelatase; ferrochelatase, mitochondrial; Heme synthase; heme synthetase; protoheme ferro-lyase; protoporphyria
Gene Symbols FECH
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human FECH (NP_0001312).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14151, 2235, 361338
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.