Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fehling's Alkaline Solution (Fehling Solution B), Ricca Chemical
SDP

Catalog No. p-3251660
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

For Determination of Reducing Sugars

Specifications

Chemical Name or Material Fehling's Alkaline Solution
Color Colorless
Name Note Fehling Solution B
CAS 1310-73-2, 6381-59-5, 7732-18-5
Grade Laboratory
Packaging Natural Poly Bottle
pH >12
Quantity 500 mL
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.