Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fehling's Alkaline Solution (Fehling Solution B), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Specifications
| Chemical Name or Material | Fehling's Alkaline Solution |
| Color | Colorless |
| Name Note | Fehling Solution B |
| CAS | 1310-73-2, 6381-59-5, 7732-18-5 |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| pH | >12 |
| Quantity | 500 mL |
| Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction