Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fehling's Solution 'B', Alkaline No. 2, Certified, LabChem™

Catalog No. p-5379547
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer™
Poly Bottle

Specifications

Chemical Name or Material Fehling's Solution 'B', Alkaline No. 2
Name Note Alkaline No. 2
CAS Max % 60.9, 34.6, 4.5
CAS 6381-59-5, 1310-73-2
Linear Formula KNaC4H4O6·4H2O
UN Number UN1824
Solubility Information Soluble in water
Identification Passes Test
Formula Weight 282.22
Grade Certified
Packaging Poly Bottle
Sensitivity Passes Test
Decomposition Information Sodium oxide; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
Color Colorless
Quantity 500 mL
Physical Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.