Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fehlings Solution 'A', Copper No. 1, Certified, 6.93% (w/v) ±0.30%, LabChem™
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer™
Poly Bottle
Specifications
Specifications
| Chemical Name or Material | Fehlings Solution 'A', Copper No. 1 |
| Name Note | Copper No. 1 |
| CAS | 7758-99-8, 7732-18-5 |
| CAS Max % | 93.07, 6.93 |
| Linear Formula | CuSO4·5H2O |
| Solubility Information | Soluble in water |
| Identification | Passes Test |
| Formula Weight | 249.68 |
| Percent Purity | 6.93% ±0.3% (w/v) |
| Grade | Certified |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction